Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCV-NS3 Antibody (1B6), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB173578 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173578 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173578 Supplier Novus Biologicals Supplier No. NBP280122AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HCV-NS3 Monoclonal antibody specifically detects HCV-NS3 in Virus samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen HCV-NS3
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 1B6
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias Nonstructural Protein 3, NS3 protease, Q91RS4
Host Species Mouse
Immunogen Recombinant HCV-NS3 serine protease domain.
Purification Method >95%, by SDS-PAGE
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.