Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HCV-NS5b Antibody (5B-12B7), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB173632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173632 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173632 Supplier Novus Biologicals Supplier No. NBP280124IR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HCV-NS5b Monoclonal antibody specifically detects HCV-NS5b in Virus samples. It is validated for Immunofluorescence

Specifications

Antigen HCV-NS5b
Applications Immunofluorescence
Classification Monoclonal
Clone 5B-12B7
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias EC:2.7.7.48, Nonstructural Protein 5B, NS5B, Q8JXY1, RNA-directed RNA polymerase
Host Species Mouse
Immunogen Recombinant HCV-NS5b RNA-dependent RNA polymerase.
Purification Method >95%, by SDS-PAGE
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 951475
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.