Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC1 Antibody (10A1), Novus Biologicals™
SDP

Catalog No. NB298496 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298496 50 μL
NB298497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298496 Supplier Novus Biologicals Supplier No. NBP32624050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HDAC1 Monoclonal antibody specifically detects HDAC1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen HDAC1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 10A1
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EC 3.5.1.98, GON-10, HD1DKFZp686H12203, histone deacetylase 1, reduced potassium dependency, yeast homolog-like 1, RPD3L1RPD3
Host Species Rabbit
Immunogen A synthesized peptide derived from Human HDAC1 [UniProt Q13547]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Loading Controls, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3065
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.