Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056340T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056340T 0.025 mg
NB10056340 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056340T Supplier Novus Biologicals Supplier No. NB10056340SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 13 publications

HDAC1 Polyclonal specifically detects HDAC1 in Human, Mouse, Rat samples. It is validated for Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Simple Western, Western Blot.

Specifications

Antigen HDAC1
Applications Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Simple Western 1:200, Immunohistochemistry 1:500. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (PMID 25648995), Immunohistochemistry-Frozen 1:10-1:500. Use reported in scientific literature (PMID 26551542), Knockout Validated
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q01101
Gene Alias EC 3.5.1.98, GON-10, HD1DKFZp686H12203, histone deacetylase 1, reduced potassium dependency, yeast homolog-like 1, RPD3L1RPD3
Gene Symbols HDAC1
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 1-5, 433-448 and 467-482 of human HDAC1 (Genbank Accession no. Q13547).
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, Epigenetics, Histone Deacetylases, Loading Controls, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3065
Test Specificity In 293, a 60 kDa band is observed.
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.