Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC10 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018222 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018222 0.025 mg
NB600223 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018222 Supplier Novus Biologicals Supplier No. NB600223SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDAC10 Polyclonal specifically detects HDAC10 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen HDAC10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Simple Western
Formulation PBS with 0.02% Sodium Azide
Gene Alias DKFZP761B039, EC 3.5.1.98, HD10, histone deacetylase 10, MGC149722
Gene Symbols HDAC10
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a portion of human HDAC 10.
Molecular Weight of Antigen 62 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Checkpoint signaling, Chromatin Research, Epigenetics, Histone Deacetylases, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 83933
Test Specificity Reacts with the human HDAC 10 protein.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.