Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC10 Rabbit anti-Human, Clone: SD08-11, Novus Biologicals™
SDP

Catalog No. NBP267801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267801 Supplier Novus Biologicals Supplier No. NBP267801

Rabbit Monoclonal Antibody

HDAC10 Monoclonal specifically detects HDAC10 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HDAC10
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SD08-11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias DKFZP761B039, EC 3.5.1.98, HD10, histone deacetylase 10, MGC149722
Gene Symbols HDAC10
Host Species Rabbit
Immunogen Synthetic peptide within Human HDAC10 aa 339-388/ 669. (SwissProt: Q969S8 Human; SwissProt: Q6P3E7 Mouse)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Breast Cancer, Cancer, Checkpoint signaling, Chromatin Research, Epigenetics, Histone Deacetylases, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 83933
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.