Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC11 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216789 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216789 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216789 Supplier Novus Biologicals Supplier No. NBP216789
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDAC11 Polyclonal specifically detects HDAC11 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen HDAC11
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown, KnockDown
Classification Polyclonal
Concentration 0.48mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Simple Western, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence, KnockDown Validated
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q96DB2
Gene Alias EC 3.5.1.98, FLJ22237, HD11, histone deacetylase 11
Gene Symbols HDAC11
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HDAC11. The exact sequence is proprietary.
Molecular Weight of Antigen 39 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Epigenetics, Histone Deacetylases
Primary or Secondary Primary
Gene ID (Entrez) 79885
Test Specificity Expected to cross react based on sequence identity: Rat (89%), Bovine (88%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.