Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500126 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500126 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500126 Supplier Novus Biologicals Supplier No. NB500126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

HDAC3 Polyclonal specifically detects HDAC3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.

Specifications

Antigen HDAC3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 12.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:2500, Immunohistochemistry-Paraffin 1:500, Microarray
Formulation 10mM PBS (pH 7.4) with 15mM Sodium Azide
Gene Accession No. O15379
Gene Alias EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45
Gene Symbols HDAC3
Host Species Rabbit
Immunogen Synthetic peptide: C-NEFYDGDHDNDKESDVEI conjugated to KLH, corresponding to amino acids 411-428 of human and chicken HDAC3.
Molecular Weight of Antigen 50 kDa
Purification Method IgG purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases
Primary or Secondary Primary
Gene ID (Entrez) 8841
Test Specificity Anti-Histone Deacetylase 3 specifically recognizes histone deacetylase 3 by immunoblotting and immunoprecipitation (50 kD).
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.