Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC3 Antibody (C.15200145), Novus Biologicals™
SDP

Catalog No. NBP259189 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259189 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259189 Supplier Novus Biologicals Supplier No. NBP259189

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

HDAC3 Monoclonal specifically detects HDAC3 in Human samples. It is validated for Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HDAC3
Applications Immunocytochemistry, Immunofluorescence, ChIP Assay
Classification Monoclonal
Clone C.15200145
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 2 ug/IP
Formulation PBS with 0.05% Sodium Azide and 0.05% ProClin 300
Gene Alias EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45
Gene Symbols HDAC3
Host Species Mouse
Immunogen The exact immunogen is propietary information.
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases
Primary or Secondary Primary
Gene ID (Entrez) 8841
Target Species Human
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.