Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216791 Supplier Novus Biologicals Supplier No. NBP216791
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDAC3 Polyclonal specifically detects HDAC3 in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen HDAC3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Chromatin Immunoprecipitation (ChIP)
Formulation PBS (pH 7), 20% Glycerol with 0.025% Proclin 300
Gene Accession No. O15379
Gene Alias EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45
Gene Symbols HDAC3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HDAC3. The exact sequence is proprietary.
Molecular Weight of Antigen 49 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases
Primary or Secondary Primary
Gene ID (Entrez) 8841
Test Specificity Human. Expected to cross react based on sequence identity: Mouse (99%), Chicken (95%), Rat (99%), Zebrafish (89%), Bovine (100%), Xenopus tropicalis (92%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.