Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC4+5+9 Antibody, Novus Biologicals™
SDP

Catalog No. NB018276 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018276 0.025 mL
NBP104967 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018276 Supplier Novus Biologicals Supplier No. NBP1049670.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDAC4+5+9 Polyclonal specifically detects HDAC4+5+9 in Human, Mouse, Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HDAC4+5+9
Applications Immunohistochemistry (Paraffin), Immunohistochemistry, Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl and 50% glycerol. with 0.02% Sodium Azide
Gene Accession No. P56524
Gene Alias HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3
Gene Symbols HDAC4
Host Species Rabbit
Immunogen The antiserum was produced against synthesized non-phosphopeptide derived from human HDAC4/HDAC5/HDAC9 around the phosphorylation site of serine 246/259/220 (T-A-Sp-E-P).
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases
Primary or Secondary Primary
Gene ID (Entrez) 9759
Test Specificity HDAC4/HDAC5/HDAC9 (Ab-246/259/220) detects endogenous levels of total HDAC4/HDAC5/HDAC9 protein.
Target Species Mouse, Human, Drosophila
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.