Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC4+5+9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $501.50
Specifications
| Antigen | HDAC4+5+9 |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Immunofluorescence, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
HDAC4+5+9 Polyclonal specifically detects HDAC4+5+9 in Human, Mouse, Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| HDAC4+5+9 | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Immunofluorescence, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Mouse, Human, Drosophila | |
| HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3 | |
| HDAC4 | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases | |
| P56524 | |
| 9759 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title