Learn More
Description
Specifications
Specifications
| Antigen | HDAC5 |
| Applications | Chemotaxis, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 - 2.0 ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 32196931), Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:100, Chromatin Immunoprecipitation (ChIP) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | Q9UQL6 |
| Gene Alias | Antigen NY-CO-9, EC 3.5.1.98, FLJ90614, HD5, histone deacetylase 5, KIAA0600, NY-CO-9 |
| Gene Symbols | HDAC5 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
