Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC5 Antibody (CL8121), Novus Biologicals™
SDP

Catalog No. NB036495 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036495 Supplier Novus Biologicals Supplier No. NBP288913
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HDAC5 Monoclonal specifically detects HDAC5 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HDAC5
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL8121
Conjugate Unconjugated
Dilution Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS, pH 7.2, containing 40% glycerol
Gene Alias Antigen NY-CO-9, EC 3.5.1.98, FLJ90614, HD5, histone deacetylase 5, KIAA0600, NY-CO-9
Host Species Mouse
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids: KSKEPTPGGLNHSLPQHPK
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 10014
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.