Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC5 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB005039 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB005039 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB005039 Supplier Novus Biologicals Supplier No. NBP222152C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDAC5 Polyclonal specifically detects HDAC5 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HDAC5
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Antigen NY-CO-9, EC 3.5.1.98, FLJ90614, HD5, histone deacetylase 5, KIAA0600, NY-CO-9
Gene Symbols HDAC5
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human HDAC5 protein (between residues 1-100) [UniProt Q9UQL6]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Phospho Specific
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.