Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC5 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB337538 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337538 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337538 Supplier Novus Biologicals Supplier No. NBP222152PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDAC5 Polyclonal antibody specifically detects HDAC5 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ChIP assay (ChIP)

Specifications

Antigen HDAC5
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP)
Formulation PBS
Gene Alias Antigen NY-CO-9, EC 3.5.1.98, FLJ90614, HD5, histone deacetylase 5, KIAA0600, NY-CO-9
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human HDAC5 protein (between residues 1-100) [UniProt Q9UQL6]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 10014
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.