Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC6 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10056343 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056343 0.1 mg
NB10056343T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056343 Supplier Novus Biologicals Supplier No. NB10056343
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse samples. It is validated for ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Simple Western, Western Blot.

Specifications

Antigen HDAC6
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 - 5 ug/ml, Simple Western 1:20, Chromatin Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Imbriano (2005)), Immunocytochemistry/Immunofluorescence 2 - 5 ug/ml, Immunoprecipitation assay dependent, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_002495
Gene Alias EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901
Gene Symbols HDAC6
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human HDAC6 (between amino acids 1-50) [UniProt Q9UBN7]
Purification Method Immunogen affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 10013
Test Specificity In NIH 3T3, a 134 kDa band is observed.
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.