Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP145611 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145611 0.1 mg
NB018570 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP145611 Supplier Novus Biologicals Supplier No. NBP145611

Rabbit Polyclonal Antibody

HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen HDAC6
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 4 ug/mL, Immunohistochemistry 20 ug/mL
Formulation PBS (pH 7.2), 5 mM EDTA, 0.5% BSA, 30% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9Z2V5
Gene Alias EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901
Gene Symbols HDAC6
Host Species Rabbit
Immunogen Synthetic peptide mapping to the N-terminus of mouse HDAC-6.
Molecular Weight of Antigen 131 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases
Primary or Secondary Primary
Gene ID (Entrez) 10013
Test Specificity The antibody does not cross-react with other HDAC proteins including HDAC1, 2, 3, 4, 5, 7, and 8.
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.