Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDAC6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
Antigen | HDAC6 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1692720
![]() |
Novus Biologicals
NBP16912720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP169127
![]() |
Novus Biologicals
NBP169127 |
100 μL |
Each for $499.50
|
|
|||||
Description
HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HDAC6 | |
Unconjugated | |
RUO | |
10013 | |
Synthetic peptides corresponding to Hdac6 (histone deacetylase 6) The peptide sequence was selected from the N terminal of Hdac6. Peptide sequence RQRKSRHNPQSPLQDSSATLKRGGKKGAVPHSSPNLAEVKKKGKMKKLSQ. | |
Primary |
Polyclonal | |
Rabbit | |
EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901 | |
HDAC6 | |
IgG | |
125 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title