Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC6 Antibody (PCRP-HDAC6-1A4), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB156660 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB156660 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB156660 Supplier Novus Biologicals Supplier No. NBP314206AF594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HDAC6 Monoclonal antibody specifically detects HDAC6 in Human samples. It is validated for Flow Cytometry, Immunofluorescence

Specifications

Antigen HDAC6
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-HDAC6-1A4
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901
Host Species Mouse
Immunogen Recombinant full-length human HDAC6 protein (Uniprot: Q9UBN7)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10013
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.