Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC6 Rabbit anti-Human, Mouse, Alexa Fluor™ 532, Polyclonal, Novus Biologicals™
SDP

Catalog No. N106343A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106343A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106343A32 Supplier Novus Biologicals Supplier No. NB10056343AF532

Rabbit Polyclonal Antibody

HDAC6 Polyclonal specifically detects HDAC6 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HDAC6
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 3.5.1.98, HD6FLJ16239, histone deacetylase 6, JM21, KIAA0901
Gene Symbols HDAC6
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human HDAC6 (between amino acids 1-50) [UniProt Q9UBN7]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10013
Test Specificity In NIH 3T3, a 134 kDa band is observed.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.