Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC7 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB055235 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB055235 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB055235 Supplier Novus Biologicals Supplier No. NBP298759C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDAC7 Polyclonal specifically detects HDAC7 in Human samples. It is validated for Western Blot.

Specifications

Antigen HDAC7
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human HDAC7.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 51564
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.