Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDAC9 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB337683 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337683 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337683 Supplier Novus Biologicals Supplier No. NBP203993F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HDAC9 Polyclonal antibody specifically detects HDAC9 in Human, Mouse, Bovine, Canine, Primate samples. It is validated for Western Blot

Specifications

Antigen HDAC9
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias EC 3.5.1.98, HD7b, HD7HDAC, HD9, HDAC7, HDAC7BDKFZp779K1053, HDRP, histone deacetylase 4/5-related protein, Histone deacetylase 7B, histone deacetylase 9, Histone deacetylase-related protein, KIAA0744HDAC9B, MEF-2 interacting transcription repressor (MITR) protein, MEF2-interacting transcription repressor MITR, MITRHDAC9FL
Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human HDAC-9 was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 9734
Target Species Human, Mouse, Bovine, Canine, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.