Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDDC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | HDDC3 |
|---|---|
| Concentration | 0.26 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3253
![]()
|
Proteintech Group Inc
210911AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
HDDC3 is involved in biological processes as a hydrolyzing enzyme in starvation response.Specifications
| HDDC3 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8N4P3, Q9D114 | |
| 308758, 374659, 68695 | |
| HDDC3 Fusion Protein Ag15342 | |
| Primary | |
| -20°C | |
| HDDC3 |
| 0.26 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| HD domain containing 3, HD domain containing protein 3, HDDC3 | |
| Hddc3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title