Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDGF Antibody (OTI8C7), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP270882R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270882R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270882R Supplier Novus Biologicals Supplier No. NBP270882R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HDGF Monoclonal specifically detects HDGF in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen HDGF
Applications Western Blot
Classification Monoclonal
Clone OTI8C7
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ96580, hepatoma-derived growth factor, hepatoma-derived growth factor (high-mobility group protein 1-like), High mobility group protein 1-like 2, HMG-1L2, HMG1L2DKFZp686J1764
Gene Symbols HDGF
Host Species Mouse
Immunogen Full length recombinant protein of human HDGF (NP_004485) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3068.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.