Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDGF Antibody (OTI8C7), PerCP, Novus Biologicals™
SDP

Catalog No. NB289283 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289283 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289283 Supplier Novus Biologicals Supplier No. NBP270882PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HDGF Monoclonal antibody specifically detects HDGF in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen HDGF
Applications Western Blot
Classification Monoclonal
Clone OTI8C7
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias FLJ96580, hepatoma-derived growth factor, hepatoma-derived growth factor (high-mobility group protein 1-like), High mobility group protein 1-like 2, HMG-1L2, HMG1L2DKFZp686J1764
Host Species Mouse
Immunogen Full length human recombinant protein of human HDGF(NP_004485) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 3068
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.