Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDGFRP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP17964520UL
Description
HDGFRP3 Polyclonal specifically detects HDGFRP3 in Human samples. It is validated for Western Blot.Specifications
HDGFRP3 | |
Polyclonal | |
Western Blot 1:1000 | |
NP_057157 | |
HDGFRP3 | |
Synthetic peptide directed towards the middle region of human HDGFRP3The immunogen for this antibody is HDGFRP3. Peptide sequence GSKRKKSYTSKKSSKQSRKSPGDEDDKDCKEEENKSSSEGGDAGNDTRNT. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
HDGF2, HDGF-2, Hepatoma-derived growth factor 2, hepatoma-derived growth factor, related protein 3, hepatoma-derived growth factor-related protein 3, HRP-3 | |
Rabbit | |
22 kDa | |
20 μL | |
Cytokine Research | |
50810 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction