Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HDGFRP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | HDGFRP3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17964520
![]() |
Novus Biologicals
NBP17964520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179645
![]() |
Novus Biologicals
NBP179645 |
100 μL |
Each for $487.50
|
|
|||||
Description
HDGFRP3 Polyclonal specifically detects HDGFRP3 in Human samples. It is validated for Western Blot.Specifications
HDGFRP3 | |
Polyclonal | |
Rabbit | |
Cytokine Research | |
HDGF2, HDGF-2, Hepatoma-derived growth factor 2, hepatoma-derived growth factor, related protein 3, hepatoma-derived growth factor-related protein 3, HRP-3 | |
HDGFRP3 | |
IgG | |
22 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_057157 | |
50810 | |
Synthetic peptide directed towards the middle region of human HDGFRP3The immunogen for this antibody is HDGFRP3. Peptide sequence GSKRKKSYTSKKSSKQSRKSPGDEDDKDCKEEENKSSSEGGDAGNDTRNT. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title