Learn More
Description
Specifications
Specifications
| Antigen | HDHD1A |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI7A2 |
| Conjugate | APC-Cyanine7 |
| Dilution | Flow Cytometry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | DXF68S1E5′-PsiMPase, EC 3.1.3.n6, family with sequence similarity 16, member A, X-linked, GS1FAM16AX, haloacid dehalogenase-like hydrolase domain containing 1, haloacid dehalogenase-like hydrolase domain containing 1A, Haloacid dehalogenase-like hydrolase domain-containing protein 1, Haloacid dehalogenase-like hydrolase domain-containing protein 1A, HDHD1Apseudouridine-5′-monophosphatase, Protein GS1 |
| Gene Symbols | PUDP |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
