Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ HDJ-2/DNAJ Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments.

Specifications

Antigen HDJ-2/DNAJ Ab-1
Clone KA2A5.6
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 4
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS225P1
View Documents
Thermo Scientific™
MS225P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS225R1
View Documents
Thermo Scientific™
MS225R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS225B
View Documents
Thermo Scientific™
MS225B
Biotinylated Ab with BSA and Azide 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS225P0
View Documents
Thermo Scientific™
MS225P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments with Thermo Scientific™ HDJ-2/DNAJ Ab-1, Mouse Monoclonal Antibody.

It is a novel human homolog of E. coli's DNAJ protein from heat shock protein 40 family. Hsp40 is involved in chaperonic protein folding HDJ-2 is cytoplasmic and nuclear membrane associated protein; upon heat shock it migrates and becomes associated with the Golgi complex, nucleolus and nuclear membrane HDJ-2 has a prenylated C-terminus and is not inducible upon heat shock.

Host Species: Mouse

Clone: KA2A5.6

Isotype: IgG1

Species Reactivity: Human, Monkey, Pig, Rabbit, Mouse, Rat, Hamster and Bacteria.

Epitope: aa 1-179

Immunogen: N-terminal purified recombinant fragment (aa 1-179) of HDJ-2 fusion protein

Molecular Weight: 44-47 kDa

Positive Control: MAD109, Raji cells, Tonsil

Cellular Localization: Cytoplasm (ground state) and Golgi complex, Nucleolus, and Nuclear membrane (upon heat shock)

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

HDJ-2/DNAJ Ab-1
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Stress Response
N-terminal purified recombinant fragment (aa 1-179) of HDJ-2 fusion protein
KA2A5.6
Monoclonal
RUO
Primary
IgG1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit