Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDJ2 Antibody (23C6), Novus Biologicals™
SDP

Catalog No. NB297744 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297744 50 μL
NB297745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297744 Supplier Novus Biologicals Supplier No. NBP32661650UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HDJ2 Monoclonal antibody specifically detects HDJ2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry

Specifications

Antigen HDJ2
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 23C6
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias dj-2, DjA1, DnaJ (Hsp40) homolog, subfamily A, member 1, dnaJ homolog subfamily A member 1, DnaJ protein homolog 2, DNAJ2, hDJ-2, HDJ2, Heat shock 40 kDa protein 4, Heat shock protein J2, heat shock protein, DNAJ-like 2, HSDJ, HSJ2, HSJ-2, HSPF4hdj-2, Human DnaJ protein 2, NEDD7, neural precursor cell expressed, developmentally down-regulated 7
Host Species Rabbit
Immunogen A synthesized peptide derived from Human HDJ2 [UniProt P31689]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline DNA Repair, Golgi Apparatus Markers, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3301
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.