Learn More
Description
Specifications
Specifications
| Antigen | HDJ2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS containing 50% glycerol |
| Gene Alias | dj-2, DjA1, DnaJ (Hsp40) homolog, subfamily A, member 1, dnaJ homolog subfamily A member 1, DnaJ protein homolog 2, DNAJ2, hDJ-2, HDJ2, Heat shock 40 kDa protein 4, Heat shock protein J2, heat shock protein, DNAJ-like 2, HSDJ, HSJ2, HSJ-2, HSPF4hdj-2, Human DnaJ protein 2, NEDD7, neural precursor cell expressed, developmentally down-regulated 7 |
| Host Species | Rabbit |
| Immunogen | Recombinant human HDJ2. |
| Purification Method | Protein A purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
