Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HE4/WFDC2 Antibody (404), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB037570 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB037570 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB037570 Supplier Novus Biologicals Supplier No. NBP290132AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HE4/WFDC2 Monoclonal specifically detects HE4/WFDC2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HE4/WFDC2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 404
Conjugate Alexa Fluor 750
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias dJ461P17.6, EDDM4, Epididymal secretory protein E4, epididymis-specific, whey-acidic protein type, four-disulfide core, HE4MGC57529, Major epididymis-specific protein E4, Putative protease inhibitor WAP5, WAP domain containing protein HE4-V4, WAP four-disulfide core domain 2, WAP four-disulfide core domain protein 2, WAP5epididymal protein 4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human HE4/WFDC2 (NP_006094.3; Met1-Phe124).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10406
Test Specificity No cross-reactivity in ELISA with: Rat WFDC2
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.