Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Hearing Loss Markers Antibody Pack
SDP

Catalog No. nbp178947 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178947 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178947 Supplier Novus Biologicals™ Supplier No. NBP178947

For Research applications

Specifications

Antigen Hearing Loss Markers
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry, Knockdown Validated
Formulation See individual datasheets.
Immunogen See individual datasheets.
Quantity 2 Vials
Research Discipline Hypoxia
Test Specificity This Hearing Loss Markers Antibodies Sample Pack includes antibodies to DUOX2 and Glutathione Peroxidase 3.
Includes 1.NB110-61576: DUOX2 Antibody
2.NBP1-06398: Glutathione Peroxidase 3/GPX3 Antibody
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.