Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heartland Virus Glycoprotein 1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB435946 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB435946 0.025 mg
NBP241228 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB435946 Supplier Novus Biologicals Supplier No. NBP2412280.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Heartland Virus Glycoprotein 1 Polyclonal specifically detects Heartland Virus Glycoprotein 1 in Virus samples. It is validated for ELISA.

Specifications

Antigen Heartland Virus Glycoprotein 1
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias Glycoprotein polyprotein
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the carboxy terminus of the Heartland virus glycoprotein 1. The immunogen is located within amino acids 520 - 570 of Heartland Virus Glycoprotein 1. Amino Acid Squence: ERGEQNWGRQGYGPRGPIR
Molecular Weight of Antigen 62 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.