Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heartland Virus Glycoprotein 2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB201955 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB201955 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB201955 Supplier Novus Biologicals Supplier No. NBP241229JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Heartland Virus Glycoprotein 2 Polyclonal antibody specifically detects Heartland Virus Glycoprotein 2 in Virus samples. It is validated for ELISA

Specifications

Antigen Heartland Virus Glycoprotein 2
Applications ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Glycoprotein polyprotein
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the center of the Heartland virus glycoprotein 2. The immunogen is located within amino acids 860 - 910 of Heartland Virus Glycoprotein 2. Amino Acid Squence: VRFDQGNHGESRIYGSPLD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.