Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heartworm Antibody (HW1-39), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB163168FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163168FR 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB163168FR Supplier Novus Biologicals Supplier No. NB10063168FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Heartworm Monoclonal specifically detects Heartworm in Invertebrate samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Heartworm
Applications ELISA, Western Blot
Classification Monoclonal
Clone HW1-39
Conjugate DyLight 680
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Dirofilaria immitus
Host Species Mouse
Immunogen Made to Invertebrates HEARTWORM
Purification Method Protein A purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-63168 recognizes both male and female ether extracted parasite antigen of canine heartworm. Canine heartworm infection, caused by the filarial nematode Dirofilaria immitis is an insidious and potentially fatal condition transmitted by mosquitoes, which primarily affects the heart and lungs of dogs, but may also affect other species including cats, foxes, sea-lions, ferrets and humans. Canine heartworm disease, resulting ultimately in damage to the pulmonary vasculature system, is largely attributed to the immunological response of the host to the presence of the adult parasites, and in many cases their microscopic offspring (microfilaria) which circulate in the bloodstream, but growing evidence suggests the involvement of further factors/mediators.
Target Species Invertebrate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.