Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments

Catalog No. AB3738
Change view
Click to view available options
Material:
Aluminum
Product Type:
Easy Pierce
Easy-Peel
EasyPierce 20 μm Thin Foil
ThermoSeal
Dimensions (L x W):
78 mm x 370 m
78 mm x 610 m
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type Dimensions (L x W)
AB-3738 Aluminum Easy Pierce 78 mm x 610 m
AB-3559 Aluminum ThermoSeal 78 mm x 370 m
AB3720 Aluminum EasyPierce 20 μm Thin Foil 78 mm x 610 m
AB-3739 Aluminum Easy-Peel 78 mm x 610 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AB-3738 Supplier Thermo Scientific™ Supplier No. AB3738
Only null left
Add to Cart
Edge
Add to Cart

Save time and costs while improving sealing integrity with these microplate heat sealing instruments. Offers the capability of retrieving samples and then re-sealing the plate.

Save time and costs while improving sealing integrity with Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments. Ideal for storage applications, heat sealing offers the capability of retrieving samples and then re-sealing the plate. When heat sealing, the sealing material is first placed in position over the plate, which must have a raised rim around each well. Heat is applied evenly for several seconds, resulting in binding of the sealing material to the raised rim to achieve a complete seal.

Features:

  • Quick operation
  • Safe for temperature-sensitive samples
  • Designed for reliable performance with Thermo Scientific ALPS instruments, including the, ALPS 3000, ALPS 25 and ALPS 50 V
TRUSTED_SUSTAINABILITY

Specifications

Color Foil
Solvent Or Alcohol Resistant Life Science Reagents Resistant, DMSO Resistant
Material Aluminum
Sterility Non-sterile
Autoclavable Not Autoclavable
No. of Wells 96
No. per Case 1
No. per Pack 1
Dimensions (L x W) 78 mm x 610 m
For Use With (Application) ideal for compound management and high throughput screening applications, suitable for PCR
Content And Storage General; Long term
Format Roll
For Use With (Equipment) ALPS 5000 & ALPS 3000
Product Type Easy Pierce
Product Line Abgene
Adhesive Polypropylene
Certified RNase/DNase-free No
Pierceable Yes
Temperature Range (Metric) -80°C to +120°C
Well Shape All well shapes
Plate Compatibility Polypropylene, Polyethylene
Incubation Conditions General
Peelable Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.