Learn More
Restek™ Heater Block Retaining Nuts
For Agilent GC Split/Splitless Injectors
$46.15 - $56.50
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
06-611-197
![]() |
Restek™
22080 |
Agilent 5890 GCs |
Each for $56.50
|
|
|||||
06-611-254
![]() |
Restek™
23042 |
Agilent 6850/6890 GCs |
Each for $46.15
|
|
|||||
Description
- Aluminum construction
- Meets or exceeds original manufacturer's performance
Specifications
Heater Block Retaining Nut | |
Aluminum | |
1 |