Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEATR8 Mouse anti-Human, Clone: A5-D4-A6, Novus Biologicals™
SDP

Catalog No. NBP266884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266884 Supplier Novus Biologicals Supplier No. NBP266884

Mouse Monoclonal Antibody

HEATR8 Monoclonal specifically detects HEATR8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HEATR8
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A5-D4-A6
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias DKFZp781B0622, HEAT repeat containing 8
Gene Symbols MROH7
Host Species Mouse
Immunogen Recombinant protein within Human HEATR8 aa 1,025-1,323/ 1,323. (SwissProt: Q68CQ1 Human)
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 374977
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.