Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HECTD1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB291357 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291357 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291357 Supplier Novus Biologicals Supplier No. NBP149926PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HECTD1 Polyclonal antibody specifically detects HECTD1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen HECTD1
Applications Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias E3 ligase for inhibin receptor, E3 ubiquitin-protein ligase HECTD1, EC 6.3.2.-, EULIR, HECT domain containing 1, HECT domain-containing protein 1, KIAA1131FLJ38315
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 2100 and 2150 of human HECT Domain Containing 1 using the numbering given in entry AAP13073.1 (GeneID 25831).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25831
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.