Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HECTD2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen HECTD2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP155081
SDP
View Documents
Novus Biologicals
NBP155081
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

HECTD2 Polyclonal specifically detects HECTD2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

HECTD2
Polyclonal
Rabbit
EC 6.3.2.-, FLJ16050, FLJ37306, HECT domain containing 2, HECT domain-containing protein 2, probable E3 ubiquitin-protein ligase HECTD2
HECTD2
IgG
88 kDa
Western Blot
Unconjugated
RUO
143279
Synthetic peptides corresponding to HECTD2(HECT domain containing 2) The peptide sequence was selected from the C terminal of HECTD2. Peptide sequence TDLTIKYFWDVVLGFPLDLQKKLLHFTTGSDRVPVGGMADLNFKISKNET.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.