Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helicobacter Pylori Antibody (HP/1334), FITC, Novus Biologicals™
SDP

Catalog No. NB179505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB179505 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB179505 Supplier Novus Biologicals Supplier No. NBP314092F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Helicobacter Pylori Monoclonal antibody specifically detects Helicobacter Pylori in H. Pylori samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen Helicobacter Pylori
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone HP/1334
Conjugate FITC
Formulation PBS
Gene Alias H. Pylori
Host Species Mouse
Immunogen Total sonicate of Helicobacter pylori
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species H. pylori
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.