Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helicobacter Pylori Antibody (HP/1335), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB387070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387070 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387070 Supplier Novus Biologicals Supplier No. NBP308426AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Helicobacter Pylori Monoclonal antibody specifically detects Helicobacter Pylori in H. pylori samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Helicobacter Pylori
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone HP/1335
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias H. pylori
Host Species Mouse
Immunogen Total sonicate of Helicobacter pylori
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species H. pylori
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.