Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helicobacter Pylori Antibody, Novus Biologicals™
SDP

Catalog No. NB018091 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018091 0.1 mL
NB1207788 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018091 Supplier Novus Biologicals Supplier No. NB12077880.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Helicobacter Pylori Polyclonal specifically detects Helicobacter Pylori in Bacteria samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Helicobacter Pylori
Applications Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation EDTA, pH 8.0, containing less than 0.1% sodium azide with Sodium Azide
Gene Alias H. pylori
Host Species Rabbit
Immunogen Total lysate of H. pylori.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This reacts with heat stable, somatic antigens of the whole H. pylori organism.
Target Species Bacteria
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.