Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helicobacter Pylori Antibody (RM411)
SDP

Catalog No. NB036686 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036686 Supplier Novus Biologicals Supplier No. NBP289101

Rabbit Monoclonal Antibody

Helicobacter Pylori Monoclonal specifically detects Helicobacter Pylori in Non-species specific samples. It is validated for ELISA.

Specifications

Antigen Helicobacter Pylori
Applications ELISA
Classification Monoclonal
Clone RM411
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA 0.05 ug/ml - 0.5 ug/ml
Formulation PBS, 50% Glycerol, 1% BSA
Gene Alias H. pylori
Host Species Rabbit
Immunogen OMP proteins purified from Helicobacter pylori
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.