Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helicobacter Pylori Mouse anti-Bacteria, Clone: HP/1336, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP30767420UG

 View more versions of this product

Catalog No. NB122822


Only null left
Add to Cart

Description

Description

Helicobacter Pylori Monoclonal specifically detects Helicobacter Pylori in Bacteria samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Helicobacter Pylori
Monoclonal
0.2 mg/ml
Flow Cytometry 1-2 ug/million cells, ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
H. pylori
Total sonicate of Helicobacter pylori
20 μg
Primary
Store at 4°C.
IgG1 κ
Flow Cytometry, ELISA, Immunofluorescence
HP/1336
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Bacteria
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.