Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helicobacter Pylori Mouse anti-Bacteria, Clone: HP/1336, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen Helicobacter Pylori
Clone HP/1336
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Flow Cytometry, ELISA, Immunofluorescence
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122822
SDP
View Documents
Novus Biologicals
NBP30767420UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

Helicobacter Pylori Monoclonal specifically detects Helicobacter Pylori in Bacteria samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Helicobacter Pylori
0.2 mg/ml
Flow Cytometry, ELISA, Immunofluorescence
Unconjugated
Mouse
Bacteria
H. pylori
IgG1 κ
Protein A or G purified
HP/1336
Flow Cytometry 1-2 ug/million cells, ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
Total sonicate of Helicobacter pylori
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.