Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helicobacter pylori urease B Antibody (1263), Novus Biologicals™
SDP

Catalog No. NBP242853 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242853 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242853 Supplier Novus Biologicals Supplier No. NBP242853
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, ELISA

Helicobacter pylori urease B Monoclonal specifically detects Helicobacter pylori urease B in Bacteria samples. It is validated for Western Blot.

Specifications

Antigen Helicobacter pylori urease B
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1263
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:20000, ELISA 1:1000-1:10000
Formulation PBS with No Preservative
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of Helicobacter pylori Urease B. The exact sequence is proprietary.
Molecular Weight of Antigen 62 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.