Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helios Antibody (22F6), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP237723IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237723IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237723IR Supplier Novus Biologicals Supplier No. NBP237723IR
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Helios Monoclonal specifically detects Helios in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Helios
Applications Flow Cytometry
Classification Monoclonal
Clone 22F6
Conjugate DyLight 755
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2
Gene Symbols IKZF2
Host Species Armenian Hamster
Immunogen Peptide coresponding to the amino acids 51-107 of Helios.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 22807
Test Specificity The armenian hamster monoclonal antibody 22F6 recognizes Helios, a transcription factor expressed in some hematopoietic stem cells, and at high levels in thymic-derived regulatory T cells.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.